Iridium ≥99.7% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36149.FF 36149.FI
36149.FFEA 58190 DKK
36149.FF 36149.FI
Iridium ≥99.7% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Iridium
Purity: 99.7+% CAS No.: 7439-88-5 Physical Form: ≈7g/25x25mm MDL No.: MFCD00011062
Formel: Ir
Molekylvægt: 192.22 g/mol
Kogepunkt: 5300
Smeltepunkt: 2450
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011062
CAS-nummer: 7439-88-5
EINECS: 231-095-9