Jern ≥99.99% (metals basis), rods, Ø 12 mm (0.47 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

40499.DJ 40499.HF
40499.DJEA 5920 DKK
40499.DJ 40499.HF
Jern ≥99.99% (metals basis), rods, Ø 12 mm (0.47 in)
Jern

Purity: 99.99% CAS No.: 7439-89-6 Physical Form: ≈0.89g/mm MDL No.: MFCD00010999

Formel: Fe
Kogepunkt: 3000 °C (1013 hPa)
Smeltepunkt: 1535 °C
Massefylde: 7,86 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010999
CAS-nummer: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR