Jern ≥99.995% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

11448.FF 11448.FI
11448.FFEA 2850 DKK
11448.FF 11448.FI
Jern ≥99.995% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Jern

Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈4.9g/25x25mm MDL No.: MFCD00010999

Formel: Fe
Molekylvægt: 55.85 g/mol
Kogepunkt: 3000
Smeltepunkt: 1535
Massefylde: 7.86 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010999
CAS-nummer: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR