Jern ≥99.995% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11381.FF 11381.FI 11381.GH
11381.FFEA 932 DKK
11381.FF 11381.FI 11381.GH
Jern ≥99.995% (metals basis), foil, Puratronic®, Thickness: 0.1 mm (0.004 in)
Jern
Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈0.49g/25x25mm MDL No.: MFCD00010999
Formel: Fe
Kogepunkt: 3000 °C (1013 hPa)
Smeltepunkt: 1535 °C
Massefylde: 7,86 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010999
CAS-nummer: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109