Jern 99,994 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

11379.FF 11379.FI 11379.GH
11379.FFEA 1670 DKK
11379.FF 11379.FI 11379.GH
Jern 99,994 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Jern

Purity: 99.994% CAS No.: 7439-89-6 Physical Form: ≈2.5g/25x25mm MDL No.: MFCD00010999

Formel: Fe
Molekylvægt: 55.85 g/mol
Kogepunkt: 3000
Smeltepunkt: 1535
Massefylde: 7.86 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010999
CAS-nummer: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR