Cobalt ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
42661.FF 42661.FI
42661.FFEA 2240 DKK
42661.FF 42661.FI
Cobalt ≥99.95% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Cobalt
Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈11.2g/25x25mm Odour: Odourless MDL No.: MFCD00010935
Formel: Co
Kogepunkt: 2870 °C (1013 hPa)
Smeltepunkt: 1495 °C
Massefylde: 8,9 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010935
CAS-nummer: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488