Mangan ≥99.6% (metals basis), powder -140+325 mesh

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

41343.14 41343.22 41343.36
41343.14EA 522 DKK
41343.14 41343.22 41343.36
Mangan ≥99.6% (metals basis), powder -140+325 mesh
Manganese

Purity: 99.6% CAS No.: 7439-96-5 UN No.: 3089 MDL No.: MFCD00011111

Formel: Mn
Molekylvægt: 54.94 g/mol
Massefylde: 7.2 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011111
CAS-nummer: 7439-96-5
EINECS: 231-105-1
UN nummer: 3089
ADR: 4.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR