Iod(I)chlorid solution 0.1 M ICl acc. Wijs volumetric solution, Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Iodochloride
35071-2.5L 35071-1L 35071-4X2.5L 35071-6X1L
35071-2.5LEA 1780 DKK
35071-2.5L 35071-1L 35071-4X2.5L 35071-6X1L
Iod(I)chlorid solution 0.1 M ICl acc. Wijs volumetric solution, Fluka™
Iodine monochloride
Iodine according to Wijs for the iodine value determination
Formel: ClI
Molekylvægt: 162.36 g/mol
Kogepunkt: 118 °C (1.013 hPa)
Smeltepunkt: 26
Massefylde: 1.06 g/cm³ (20 °C)
Flammepunkt: 40 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011354
CAS-nummer: 7790-99-0
UN nummer: 2789
ADR: 8 II
Merck Index: 13,05039
Bestil nu
Specifikationer Testresultater
Factor 0.99 - 1.01