Contact slides SDX, yeasts and moulds, HYCON®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.44017.0020 1.44016.0020
BTSM1.44017.0020EA 950 DKK
BTSM1.44017.0020 BTSM1.44016.0020
Contact slides SDX, yeasts and moulds, HYCON®
Mikrobiologiske medier Klargjort medium
Sabouraud dextrose agar with modified Pharmacopoeia formulation for the determination of yeast and moulds.
Contains neutralisers (Tween® 80, L-a-Phosphatidylcholine) and growth supplements.