Vanadium ≥99.5% (metals basis), folie, Thickness: 1.0 mm (0.04 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11097.FF 11097.FI 11097.GA
11097.FFEA 1410 DKK
11097.FF 11097.FI 11097.GA
Vanadium ≥99.5% (metals basis), folie, Thickness: 1.0 mm (0.04 in)
Vanadium
Purity: 99.5% CAS No.: 7440-62-2 Physical Form: ≈3.8g/25x25mm Odour: Odourless MDL No.: MFCD00011453
Formel: V
Kogepunkt: 3477 °C (1013 hPa)
Smeltepunkt: 1896 °C
Massefylde: 6,015 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011453
CAS-nummer: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053