Niobium ≥99.8% (metals basis), pieces
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
00241.22
00241.36
00241.22EA
1320
DKK
00241.22
00241.36
Niobium ≥99.8% (metals basis), pieces
Niobium
Purity: 99.8%
CAS No.: 7440-03-1
Appearance: Grey or silver
Odour: Odourless
MDL No.: MFCD00011126
Formel:
Nb Molekylvægt: 92.91 g/mol Kogepunkt: 3700 Smeltepunkt: 2468 Massefylde: 8.57 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011126 CAS-nummer: 7440-03-1 EINECS: 231-113-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...