Titan 99,6 Metals basis, foil, annealed, Thickness: 0.05 mm (0.002 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
44202.FI 44202.GH 44202.GT
44202.FIEA 1700 DKK
44202.FI 44202.GH 44202.GT
Titan 99,6 Metals basis, foil, annealed, Thickness: 0.05 mm (0.002 in)
Titanium
Purity: 99.6% CAS No.: 7440-32-6 Physical Form: ≈0.14g/25x25mm, Temper:soft Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
Molekylvægt: 47.88 g/mol
Kogepunkt: 3262
Smeltepunkt: 1660
Massefylde: 4.51 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011264
CAS-nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610