Titan ≥99.995% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
42393.06 42393.14
42393.06EA 811 DKK
42393.06 42393.14
Titan ≥99.995% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Titanium
Purity: 99.995% CAS No.: 7440-32-6 Physical Form: ≈0.23g/slug Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
Kogepunkt: 3262 °C (1013 hPa)
Smeltepunkt: 1660 °C
Massefylde: 4,51 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011264
CAS-nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610