Titan 99,99 Metals basis, foil, Thickness: 0.25 mm (0.01 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
13975.FF 13975.FI 13975.GH
13975.FFEA 1370 DKK
13975.FF 13975.FI 13975.GH
Titan 99,99 Metals basis, foil, Thickness: 0.25 mm (0.01 in)
Titanium
Purity: 99.99+% CAS No.: 7440-32-6 Physical Form: ≈0.7g/25x25mm Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
Molekylvægt: 47.88 g/mol
Kogepunkt: 3262
Smeltepunkt: 1660
Massefylde: 4.51 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011264
CAS-nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610