Titan 99,5 Metals basis, plate 100x100 mm (3.9x3.9 in), annealed, Thickness: 3.175 mm (0.125 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
12598.KG 12598.KJ
12598.KGEA 1610 DKK
12598.KG 12598.KJ
Titan 99,5 Metals basis, plate 100x100 mm (3.9x3.9 in), annealed, Thickness: 3.175 mm (0.125 in)
Titanium
Purity: 99.5% CAS No.: 7440-32-6 Physical Form: ≈143g/pc Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
Molekylvægt: 47.88 g/mol
Kogepunkt: 3262
Smeltepunkt: 1660
Massefylde: 4.51 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011264
CAS-nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610