Titan ≥99.99% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

10388.BU 10388.BY 10388.CC
10388.BUEA 904 DKK
10388.BU 10388.BY 10388.CC
Titan ≥99.99% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Titanium

Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈3.54g/m Odour: Odourless MDL No.: MFCD00011264

Formel: Ti
Molekylvægt: 47.88 g/mol
Kogepunkt: 3262
Smeltepunkt: 1660
Massefylde: 4.51 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011264
CAS-nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR