Titan ≥99.94% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
00360.FH
00360.FL
00360.GT
00360.FHEA
1230
DKK
00360.FH
00360.FL
00360.GT
Titan ≥99.94% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Titanium
Purity: 99.94%
CAS No.: 7440-32-6
Physical Form: ≈0.2g/25x75mm
Odour: Odourless
MDL No.: MFCD00011264
Formel:
Ti Molekylvægt: 47.88 g/mol Kogepunkt: 3262 Smeltepunkt: 1660 Massefylde: 4.51 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011264 CAS-nummer: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...