Solvent pumps, foot operated
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5603-1100 5603-3000
223-0062EA 10680 DKK
223-0062 223-0063
Solvent pumps, foot operated
Pumper Tromlepumper
Foot operated solvent pumps for filling of high purity liquids such as solvents. All medium-contacting parts are made of stainless steel AISI 304 (1.4301) and PTFE.
  • Immersion tube can be steplessly adjusted to the barrel depth
  • For barrels up to 220 litres
  • Suitable for steel barrels with R 2" thread
Due to the barrel screw joint and the stopcock, the container and the pump can be hermetically sealed. So that no dangerous vapours can escape after dispensing. This protects both health and the environment.
The solid, durable industrial version of theMore Product Information