Sølv hard ≥99.998% (metals basis), foil, Premion®, hard, Thickness: 0.25 mm (0.01 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12125.FI 12125.GH 12125.HP
12125.FIEA 1570 DKK
12125.FI 12125.GH 12125.HP
Sølv hard ≥99.998% (metals basis), foil, Premion®, hard, Thickness: 0.25 mm (0.01 in)
Sølv
Purity: 99.998% CAS No.: 7440-22-4 Physical Form: ≈6.6g/50x50mm, Temper: as rolled Odour: Characteristic MDL No.: MFCD00003397
Formel: Ag
Molekylvægt: 107.87 g/mol
Kogepunkt: 2212
Smeltepunkt: 962
Massefylde: 10.5 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003397
CAS-nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647