Myresyre 98-100% (by alkalimetry), LiChropur™ for HPLC, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Aminic Acid, Methanoic acid

M-Clarity™ quality level = MQ100

5.43804.0100 5.43804.0250
5.43804.0100EA 998 DKK
5.43804.0100 5.43804.0250
Myresyre 98-100% (by alkalimetry), LiChropur™ for HPLC, Supelco®
Myresyre
Formel: HCO₂H
Molekylvægt: 46.03 g/mol
Kogepunkt: 101 °C (1013 hPa)
Smeltepunkt: 4 °C
Massefylde: 1.22 g/cm³ (20 °C)
Flammepunkt: 49.5 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003297
CAS-nummer: 64-18-6
EINECS: 200-579-1
UN nummer: 1779
ADR: 8 (3) II

Bestil nu

Specifikationer Testresultater

Explosion Limit (V) 12 - 38%
pH Value, (10 g/l, H₂O, 20 °C) 2.2
Vapor Pressure (20 °C) 42 hPa


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR