Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

13964.FF 13964.FI
13964.FFEA 2050 DKK
13964.FF 13964.FI
Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Neodymium

Purity: 99.9% CAS No.: 7440-00-8 Physical Form: ≈0.43g/25x25mm, Packaged in mylar under argon Odour: Odourless UN No.: 3178 MDL No.: MFCD00011130

Formel: Nd
Molekylvægt: 144.24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011130
CAS-nummer: 7440-00-8
EINECS: 231-109-3
UN nummer: 3178
ADR: 4.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR