Dysprosium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

40286.DJ 40286.HF
40286.DJEA 14420 DKK
40286.DJ 40286.HF
Dysprosium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Dysprosium

Purity: 99.9% CAS No.: 7429-91-6 Molecular Formula: Packaged in mylar under argon Physical Form: ≈1.1g/mm, Cast rod, diameter is approximate Odour: Odourless

Formel: Dy
Molekylvægt: 162.5 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010982
CAS-nummer: 7429-91-6
EINECS: 231-073-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR