Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Packaged under argon.
00106.09
00106.18
00106.09EA
1800
DKK
00106.09
00106.18
Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces
Dysprosium
Purity: 99.9%
CAS No.: 7429-91-6
Physical Form: Packaged under argon
Appearance: Silver-grey
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00010982
Formel:
Dy Molekylvægt: 162.5 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010982 CAS-nummer: 7429-91-6 EINECS: 231-073-9 UN nummer: 3178 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...