Arsen ≥99.999% (metals basis), lumps

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Arsenic solution

Ampuled under argon

00590.14 00590.22
00590.14EA 2550 DKK
00590.14 00590.22
Arsen ≥99.999% (metals basis), lumps
Arsenic

Purity: 99.999% CAS No.: 7440-38-2 Physical Form: Ampouled under argon Appearance: Silver to black Odour: Odourless UN No.: 1558 MDL No.: MFCD00085309

Formel: As
Molekylvægt: 74.92 g/mol
Smeltepunkt: 817
Massefylde: 5.727 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00085309
CAS-nummer: 7440-38-2
EINECS: 213-148-6
UN nummer: 1558
ADR: 6.1 II
Merck Index: 12,00832

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR