Tin hard ≥99.9% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10789.VA 10789.VE
10789.VAEA 2470 DKK
10789.VA 10789.VE
Tin hard ≥99.9% (metals basis), foil, hard, Thickness: 0.025 mm (0.001 in)
Tin
Purity: 99.9% CAS No.: 7440-31-5 Physical Form: ≈4.2g/15x15cm, Temper: as rolled Odour: Odourless MDL No.: MFCD00133862
Formel: Sn
Molekylvægt: 118.71 g/mol
Kogepunkt: 2270
Smeltepunkt: 232
Massefylde: 7.31 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133862
CAS-nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587