Aluminium, powder

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Aluminum

5285.1 5285.2
ROTH5285.1EA 0 DKK
ROTH5285.1 ROTH5285.2
Aluminium, powder
Aluminium
Formel: Al
Kogepunkt: 2057 °C (1013 hPa)
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
UN nummer: 1396
ADR: 4.3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR