Aluminium ≥99.999% (metals basis), tråd, Puratronic®, hard, Ø 0.152 mm (0.006 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
44579.G5 44579.G9
44579.G5EA 1140 DKK
44579.G5 44579.G9
Aluminium ≥99.999% (metals basis), tråd, Puratronic®, hard, Ø 0.152 mm (0.006 in)
Aluminium
Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈0.049g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kogepunkt: 2327 °C (1013 hPa)
Smeltepunkt: 660 °C
Massefylde: 2,7 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321