Aluminium ≥99.999% (metals basis), wire, Puratronic®, hard, Ø 0.1 mm (0.004 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aluminum

44578.G5 44578.G9
44578.G5EA 1270 DKK
44578.G5 44578.G9
Aluminium ≥99.999% (metals basis), wire, Puratronic®, hard, Ø 0.1 mm (0.004 in)
Aluminium

Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈0.022g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00134029

Formel: Al
Kogepunkt: 2327 °C (1013 hPa)
Smeltepunkt: 660 °C
Massefylde: 2,7 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR