Aluminium ≥99.99% (metals basis), foil, annealed, Thickness: 1.5 mm (0.06 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Aluminum
44491.FI 44491.GH 44491.HP
44491.FIEA 1040 DKK
44491.FI 44491.GH 44491.HP
Aluminium ≥99.99% (metals basis), foil, annealed, Thickness: 1.5 mm (0.06 in)
Aluminium
Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈10.125g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kogepunkt: 2327 °C (1013 hPa)
Smeltepunkt: 660 °C
Massefylde: 2,7 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321