Aluminium ≥99.9995% (metals basis), wire, Puratronic®, annealed, Ø 0.5 mm (0.02 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Aluminum
43426.G6
43426.H2
43426.H7
43426.G6EA
1110
DKK
43426.G6
43426.H2
43426.H7
Aluminium ≥99.9995% (metals basis), wire, Puratronic®, annealed, Ø 0.5 mm (0.02 in)
Aluminium
Purity: 99.9995%
CAS No.: 7429-90-5
Physical Form: ≈0.53g/m, Temper: soft
Odour: Odourless
MDL No.: MFCD00134029
Formel:
Al Kogepunkt: 2327 °C (1013 hPa) Smeltepunkt: 660 °C Massefylde: 2,7 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00134029 CAS-nummer: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...