Aluminium ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.5 mm (0.02 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Aluminum
43425.GH 43425.HP 43425.FI
43425.GHEA 3890 DKK
43425.GH 43425.HP 43425.FI
Aluminium ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.5 mm (0.02 in)
Aluminium
Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈3.4g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Molekylvægt: 26.98 g/mol
Kogepunkt: 2327
Smeltepunkt: 660
Massefylde: 2.7 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321