Aluminium ≥99.99% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Aluminum
42328.18 42328.30
42328.18EA 771 DKK
42328.18 42328.30
Aluminium ≥99.99% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Aluminium
Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈1.1g/slug Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kogepunkt: 2327 °C (1013 hPa)
Smeltepunkt: 660 °C
Massefylde: 2,7 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321