Aluminium ≥99.9995% (metals basis), foil, Puratronic®, Thickness: 0.038 +/- 0.007 mm (0.0015 +/- 0.0003 in)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Aluminum
38366.GA 38366.HC
38366.GAEA 2280 DKK
38366.GA 38366.HC
Aluminium ≥99.9995% (metals basis), foil, Puratronic®, Thickness: 0.038 +/- 0.007 mm (0.0015 +/- 0.0003 in)
Aluminium
Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈1.03g/50x200mm Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Molekylvægt: 26.98 g/mol
Kogepunkt: 2327
Smeltepunkt: 660
Massefylde: 2.7 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321