Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aluminum

11374.FI 11374.GH 11374.HP
11374.FIEA 886 DKK
11374.FI 11374.GH 11374.HP
Aluminium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Aluminium

Purity: 99.998% CAS No.: 7429-90-5 Physical Form: ≈3.4g/50x50mm, Temper: as rolled Odour: Odourless MDL No.: MFCD00134029

Formel: Al
Molekylvægt: 26.98 g/mol
Kogepunkt: 2327
Smeltepunkt: 660
Massefylde: 2.7 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR