Aluminium ≥99.5% (metals basis), klumper
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10092.30
10092.A1
10092.A7
10092.30EA
681
DKK
10092.30
10092.A1
10092.A7
Aluminium ≥99.5% (metals basis), klumper
Aluminium
Purity: 99.5%
CAS No.: 7429-90-5
Physical Form: ≈17g/lump
Odour: Odourless
MDL No.: MFCD00134029
Formel:
Al Kogepunkt: 2327 °C (1013 hPa) Smeltepunkt: 660 °C Massefylde: 2,7 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00134029 CAS-nummer: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...