Automatic Burettes, Pellet Pattern, Class AS
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
243172704 243173306 243173606 243172753 243173355 243173655
SCOT243172704EA 2870 DKK
SCOT243172704 SCOT243173306 SCOT243173606 SCOT243172753 SCOT243173355 SCOT243173655
Automatic Burettes, Pellet Pattern, Class AS
Buretter
Burette, automatic, Pellet, DURAN® borosilicate glass, with PTFE key, class AS.
  • 30 seconds waiting time
  • Schellbach stripe and main graduations as circular divisions
  • Supplied with reservoir bottle and blow ball
Certificeringer: DIN EN ISO 385