Standards and test mixes for liquid chromatography
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31805 31800 31801 31802 31803 31804
REST31805EA 1280 DKK
REST31805 REST31800 REST31801 REST31802 REST31803 REST31804
Standards and test mixes for liquid chromatography
Standarder Kromatografi Standarder
Linear detector responses to concentration variations are an important part of operation qualification (OQ) for LC instruments. Kit of five aqueous solutions of caffeine can be used to generate simple plots of UV response versus concentration.
  • Simple check of a detector’s ability to produce a linear response to varied concentrations
  • Suitable for clear indication of linear range
Certificeringer: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.