Strips of 12 PCR tubes with detached cap strips
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
781280 781290
BRND781280EA 1720 DKK
BRND781280 BRND781290
Strips of 12 PCR tubes with detached cap strips
Rør PCR rør
PP, clear.
  • DNase, DNA and RNase-free
Separate domed lids, available in strips of 12, ensure a secure seal.
Certificeringer: DIN EN 285