Cannon-Fenske viscometers, KIMBLE®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SP46460-100 SP46460-150 SP46460-200 SP46460-300 SP46460-350 SP46460-400 SP46460-50
SCERSP46460-100EA 3190 DKK
SCERSP46460-100 SCERSP46460-150 SCERSP46460-200 SCERSP46460-300 SCERSP46460-350 SCERSP46460-400 SCERSP46460-50
Cannon-Fenske viscometers, KIMBLE®
Viskosimetre Kapillærviskosimetere
This viscometer requires calibration against a standard liquid of known viscosity or against a second viscometer with known constant.
  • Permanently marked with individual serial number
  • Lines and legend are printed black
  • Viscosity ranges shown for an efflux time greater than 200 seconds
Cannon-Fenske viscometers for kinematic viscosities of transparent liquids.
Certificeringer: ASTM D445 designed from ASTM D446