Beakers, tall form, with spout
Leverandør: QUARTZTECHNISCHE WERKSTATTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1229 1226
213-1140EA 2840 DKK
213-1140 QUTW1226
Beakers, tall form, with spout
Bægre
Quartz glass.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.
Certificeringer: DIN 12330.