pH electrodes, Sentix® IDS
Leverandør: WTW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
103740 103780 103750 103741
662-1249EA 2530 DKK
662-1249 662-1254 662-1253 662-1250
pH electrodes, Sentix® IDS
Elektroder pH-elektroder
Forbedret sensorteknologi af høj kvalitet kombineret med den mest moderne måleelektronik.
  • Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • Measurement integrity benchmarking of the electrodes by QSC (Quality Sensor Control)
  • IDS electrodes store their serial number and their calibration records
Ø×L: 12×120 mm
Bestillingsoplysninger: Forskellige kabellængder tilgængelige.