Ductless fume hoods, Chemcap™ Clearview™
Leverandør: BIGNEAT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BC0603-4 BC1206-4 BC1506-4
BIGNBC0603-4EA 62280 DKK
BIGNBC0603-4 BIGNBC1206-4 BIGNBC1506-4
Ductless fume hoods, Chemcap™ Clearview™
Stinkskabe og kabinetter Stinkskab uden Ventilation
Chemcap™ Clearview™ ductless fume hoods include Chemcap™ carbon filtration. A range of six standard cabinet sizes with keypad control panel, touch button controls and service due indicator. Includes 18 W fluorescent lamp and PVC base tray which offers excellent chemical resistance and contains spillages.
Cabinets are manufactured from polyester coated steel; transparent panels are acrylic and resistant to corrosion.
  • Fully portable
  • Clearview all-round vision
  • BC 4002 has a full width envelope access
  • BC 6003 to BC 1204 have teardrop front working access
  • BC 1206 and BC 1506 cabinets have double fan, double filter stack and have trapezium front working access
Operator protection: Chemcap™ fume cabinets protect the operator from fumes, aerosols and/or particulates whilst undertaking laboratory procedures.
Chemcap™ carbon/HEPA filtration options: Filters are included - most frequently used MP (multipurpose) and OS (where organic fumes predominate) are listed below, with or without HEPA filtration. Alternative filter types H+ (acids), F (formaldehyde), AM (ammonia) are available on request.
Power supply: 230 V, AC, 50 Hz, 5 Amp, 1 Ø (on request 110 V, AC, 60 Hz, 8 Amp, 1 Ø).
Certificeringer: Chemcap™ Clearview™ conforms to the requirements of all major standards for containment and filtration: US Standard ASHRAE 110: 1995 12-203 (for containment)
British Standard BS 7989:2001 (for filter efficiency and retention capacity). Optional rear baffle is required, please contact Avantor for details.
French Standard AFNOR NFX 15-211:2009 - Class 1 (for filter retention capacity)
Bestillingsoplysninger: Please contact Avantor for details on alternative filter types and other options, including mobile stands and service options.