Z-VAD-FMK
Leverandør: ENZO LIFE SCIENCES
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ALX260138R020
ALX260138R100
ENZOALX260138R020EA
2100
DKK
ENZOALX260138R020
ENZOALX260138R100
Z-VAD-FMK
Proteiner
Caspase inhibitor
Synthetic peptide that irreversibly inhibits ICE-family protease/caspase activity and blocks apoptosis. The inhibitor is designed as a methyl ester to facilitate cell permeability.