Ac-YVAD-CMK
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ALX260028M001 ALX260028M005
ENZOALX260028M001EA 1240 DKK
ENZOALX260028M001 ENZOALX260028M005
Ac-YVAD-CMK
Proteiner
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4. Inhibits Fas-mediated apoptosis.