Ac-YVAD-2,6-dimethylbenzoyloxymethylketone
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ALX260016M001 ALX260016M005
ENZOALX260016M001EA 1010 DKK
ENZOALX260016M001 ENZOALX260016M005
Ac-YVAD-2,6-dimethylbenzoyloxymethylketone
Proteiner
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4.