Human big endothelin-1 (1-38)

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ALX152001P001 ALX152001PC01 ALX152001PC05
ENZOALX152001P001EA 9110 DKK
ENZOALX152001P001 ENZOALX152001PC01 ENZOALX152001PC05
Human big endothelin-1 (1-38)
Proteiner

Endothelin-converting enzyme substrate


Biological precursor of endothelin 1. Cleaved to yield endothelin-1 via the activity of the endothelin-converting enzyme (ECE). Endothelins are endothelium-derived vasoconstrictor peptides.


Bestillingsoplysninger: Quantity: *Sold as net peptide weight.

Bestil nu

Specifikationer

Protein/peptide name Big Endothelin-1 (1-38) Big Endothelin-1 (1-38)
Environmentally Preferable
Species Human Human
Kilde
Renhed >98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR