Anti-HSP25 + HSP27 Rabbit Polyclonal Antibody

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ADISPA801D ADISPA801F ADISPA801J
ENZOADISPA801DEA 1680 DKK
ENZOADISPA801D ENZOADISPA801F ENZOADISPA801J
Anti-HSP25 + HSP27 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSP25 + HSP27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Pig, Hamster, Guinea pig, Gerbil, Dog, Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR