Anti-HERP Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BMLPW97050025 BMLPW97050100
ENZOBMLPW97050025EA 1180 DKK
ENZOBMLPW97050025 ENZOBMLPW97050100
Anti-HERP Rabbit Polyclonal Antibody
Antistoffer
Homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domain member 1 protein (HERP) is a membrane-associated ER protein strongly up-regulated by inducers of ER stress.
  • For Immunoprecipitation, Western Blot
Recognizes human HERP.
Type: Primary
Antigen: HERP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human