Anti-CYLD Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BMLPW07600025 BMLPW07600100
ENZOBMLPW07600025EA 799 DKK
ENZOBMLPW07600025 ENZOBMLPW07600100
Anti-CYLD Rabbit Polyclonal Antibody
Antistoffer
The tumor suppressor CYLD is a deubiquitinylating enzyme (DUB) that antagonises NF-κB and JNK signaling by disassembly of Lys63-linked ubiquitin chains synthesised in response to cytokine stimulation. The DUB activity of CYLD is mediated by its C-terminal ubiquitin specific protease (USP) domain, pathogenic truncations and mutations of which are associated with the hypertrophic skin tumor cylindromatosis through disruption of the USP domain and subsequent loss of CYLD catalytic activity.
  • For Western Blot
Recognizes human CYLD.
Type: Primary
Antigen: CYLD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu