Anti-CaN Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ADISPA610D ADISPA610F
ENZOADISPA610DEA 1680 DKK
ENZOADISPA610D ENZOADISPA610F
Anti-CaN Rabbit Polyclonal Antibody
Antistoffer
Calcineurin is a Ca2+/Calmodulin-dependent serine/threonine protein phosphatase. The active enzyme is a heterodimer consisting of a catalytic A subunit and a myristylated regulatory B subunit that binds calcium. Calcineurin, which is the target of immunosuppressant drugs such as cyclosporin A and FK506, controls the translocation of Nuclear factors of activated T-cells (NFAT), a family of transcriptional activators that control the expression of cytokine genes essential to immune response, from the cytosol into the nucleus of activated T-cells.
  • For Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, hamster, monkey, pig, rabbit and sheep calcineurin A. Detects a band of ~61kDa by Western blot.
Type: Primary
Antigen: CaN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human